• USA Home
  • 95947 - 3-Piperazinopropionitrile

EMAIL THIS PAGE TO A FRIEND
95947 Sigma-Aldrich

3-Piperazinopropionitrile

≥96.0%

Synonym: 1-(2-Cyanoethyl)piperazine

Purchase

Properties

Related Categories Building Blocks, C4 to C8, Chemical Synthesis, Heterocyclic Building Blocks, Piperazines More...
assay   ≥96.0%
impurities   ≤0.5% water
refractive index   n20/D 1.493
SMILES string   N#CCCN1CCNCC1
InChI   1S/C7H13N3/c8-2-1-5-10-6-3-9-4-7-10/h9H,1,3-7H2
InChI key   MVOFPBMQTXKONX-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?