EMAIL THIS PAGE TO A FRIEND
95980 Sigma-Aldrich

Z-Asp(OBzl)-OH

≥98.0%

Synonym: N-Cbz-L-aspartic acid β-benzyl ester, Z-L-aspartic acid 4-benzyl ester

  • CAS Number 3479-47-8

  • Empirical Formula (Hill Notation) C19H19NO6

  • Molecular Weight 357.36

  •  Beilstein/REAXYS Number 2065292

  •  EC Number 222-456-1

  •  MDL number MFCD00037820

  •  eCl@ss 32160406

  •  PubChem Substance ID 57653718

  •  NACRES NA.22

Purchase

Properties

Related Categories Amino Acids & Derivatives, Aspartic Acid, Chemical Biology, Chemical Synthesis, Peptide Synthesis and Peptide Chemistry More...
assay   ≥98.0%
SMILES string   OC(=O)[C@H](CC(=O)OCc1ccccc1)NC(=O)OCc2ccccc2
InChI   1S/C19H19NO6/c21-17(25-12-14-7-3-1-4-8-14)11-16(18(22)23)20-19(24)26-13-15-9-5-2-6-10-15/h1-10,16H,11-13H2,(H,20,24)(H,22,23)/t16-/m0/s1
InChI key   VUKCNAATVIWRTF-INIZCTEOSA-N

Description

Packaging

5 g in poly bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?