EMAIL THIS PAGE TO A FRIEND
96014 Aldrich

Z-Cys(Bzl)-ONp

≥98.0% (HPLC)

Synonym: Nα-Z-S-benzyl-L-cysteine 4-nitrophenyl ester

  • CAS Number 3401-37-4

  • Empirical Formula (Hill Notation) C24H22N2O6S

  • Molecular Weight 466.51

  •  Beilstein/REAXYS Number 1898096

  •  MDL number MFCD00024660

  •  PubChem Substance ID 24890236

Purchase

Properties

assay   ≥98.0% (HPLC)
storage temp.   2-8°C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?