EMAIL THIS PAGE TO A FRIEND
96715 Sigma-Aldrich

Z-DL-Leu-OH

≥98.0%

Synonym: Z-DL-leucine

  • CAS Number 3588-60-1

  • Empirical Formula (Hill Notation) C14H19NO4

  • Molecular Weight 265.30

  •  Beilstein Registry Number 4695869

  •  MDL number MFCD00065129

  •  PubChem Substance ID 57653777

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Leucine, Peptide Chemistry | Peptide Synthesis More...
InChI Key   USPFMEKVPDBMCG-UHFFFAOYSA-N
assay   ≥98.0%
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?