EMAIL THIS PAGE TO A FRIEND
96893 Sigma-Aldrich

Z-Lys(Z)-ONp

≥96.0% (TLC)

Synonym: Nα,Nε-di-Z-L-lysine 4-nitrophenyl ester

  • CAS Number 21160-82-7

  • Empirical Formula (Hill Notation) C28H29N3O8

  • Molecular Weight 535.55

  •  Beilstein/REAXYS Number 2319702

  •  EC Number 244-251-6

  •  MDL number MFCD00024657

  •  PubChem Substance ID 57653792

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Lysine, Peptide Synthesis and Peptide Chemistry More...
assay   ≥96.0% (TLC)
application(s)   peptide synthesis: suitable
storage temp.   2-8°C
SMILES string   [O-][N+](=O)c1ccc(OC(=O)[C@H](CCCCNC(=O)OCc2ccccc2)NC(=O)OCc3ccccc3)cc1
InChI   1S/C28H29N3O8/c32-26(39-24-16-14-23(15-17-24)31(35)36)25(30-28(34)38-20-22-11-5-2-6-12-22)13-7-8-18-29-27(33)37-19-21-9-3-1-4-10-21/h1-6,9-12,14-17,25H,7-8,13,18-20H2,(H,29,33)(H,30,34)/t25-/m0/s1
InChI key   DQKARZJTYUBTMX-VWLOTQADSA-N

Description

Packaging

5 g in poly bottle

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?