EMAIL THIS PAGE TO A FRIEND
98920 Sigma-Aldrich

Boc-D-Cys(Bzl)-OH

≥99.0%

Synonym: Boc-S-benzyl-D-cysteine

  • CAS Number 102830-49-9

  • Empirical Formula (Hill Notation) C15H21NO4S

  • Molecular Weight 311.40

  •  Beilstein Registry Number 5758339

  •  MDL number MFCD00038253

  •  PubChem Substance ID 57653813

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Cysteine/Cystine, Peptide Chemistry | Peptide Synthesis More...
InChI Key   IFVORPLRHYROAA-GFCCVEGCSA-N
assay   ≥99.0%
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?