• USA Home
  • A17900 - N-Acetyl-DL-methionine

EMAIL THIS PAGE TO A FRIEND
A17900 Sigma-Aldrich

N-Acetyl-DL-methionine

ReagentPlus®, 99%

Synonym: Methionamine

  • CAS Number 1115-47-5

  • Linear Formula CH3SCH2CH2CH(NHCOCH3)CO2H

  • Molecular Weight 191.25

  •  Beilstein/REAXYS Number 1867673

  •  EC Number 214-224-3

  •  MDL number MFCD00008681

  •  eCl@ss 32160406

  •  PubChem Substance ID 24890587

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Methionine, Peptide Synthesis and Peptide Chemistry More...
grade   ReagentPlus®
InChI Key   XUYPXLNMDZIRQH-UHFFFAOYSA-N
assay   99%
mp   117-119 °C (lit.)

Description

Packaging

250 g in glass bottle

Legal Information

ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

HPLC Analysis of N-Acetylmethionine Enantiomers on Astec® (R,R) P-CAP™

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of N-Acetylmethionine Enantiomers on Astec® (R,R) P-CAP™
Keywords: Chromatography, High performance liquid chromatography

MSMLS Online Plate Map

Metabolite descriptors included with the software download upon purchase of the product include: Name, Parent CID, molecular formula, molecular weight, CAS, ChEBI, HMDB ID, PubChem Compound and Subst...
Keywords: Mass spectrometry, Metabolites

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?