• USA Home
  • A5013 - 4-Acetylphenyl sulfate potassium salt

EMAIL THIS PAGE TO A FRIEND
A5013 Sigma-Aldrich

4-Acetylphenyl sulfate potassium salt

Purchase

Properties

storage temp.   −20°C
SMILES string   [K].CC(=O)c1ccc(OS(O)(=O)=O)cc1

Description

Substrates

A specific substrate for rat arylsulfatase C.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?