EMAIL THIS PAGE TO A FRIEND
A6316 Sigma-Aldrich

Alloxan tetrahydrate

98%

Synonym: 2,4,5,6(1H,3H)-Pyrimidinetetrone, 2,4,5,6-Tetraoxypyrimidine, 5,6-Dioxyuracil

  • CAS Number 6010-91-9

  • Empirical Formula (Hill Notation) C4H2N2O4 · 4H2O

  • Molecular Weight 214.13

  •  Beilstein/REAXYS Number 132451

  •  MDL number MFCD00149400

  •  PubChem Substance ID 57653872

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Building Blocks, C4 to C5, Chemical Synthesis,
InChI Key   JJROYYCRHFNFNC-UHFFFAOYSA-N
assay   98%
storage temp.   2-8°C
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Titrette from Brand
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?