EMAIL THIS PAGE TO A FRIEND
A66405 Sigma-Aldrich

2-Naphthylamine

95%

Synonym: β-Naphthylamine, 2-Aminonaphthalene

  • CAS Number 91-59-8

  • Linear Formula C10H7NH2

  • Molecular Weight 143.19

  •  Beilstein/REAXYS Number 3939429

  •  EC Number 202-080-4

  •  MDL number MFCD00004112

Purchase

Properties

InChI Key   JBIJLHTVPXGSAM-UHFFFAOYSA-N
assay   95%
bp   306 °C(lit.)
mp   111-113 °C (lit.)
density   1.061 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1650 6.1 / PGII
WGK Germany 
3
RTECS 
QM2100000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

N8381 2-Naphthylamine, powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?