• USA Home
  • A81609 - 3-Amino-1,2,4-triazole

EMAIL THIS PAGE TO A FRIEND
A81609 Sigma-Aldrich

3-Amino-1,2,4-triazole

95%

Synonym: 1,2,4-Triazol-3-amine, 3-AT, Amitrol

  • CAS Number 61-82-5

  • Empirical Formula (Hill Notation) C2H4N4

  • Molecular Weight 84.08

  •  Beilstein/REAXYS Number 107687

  •  EC Number 200-521-5

  •  MDL number MFCD00005230

  •  PubChem Substance ID 329770948

Purchase

Properties

InChI Key   KLSJWNVTNUYHDU-UHFFFAOYSA-N
assay   95%
mp   150-153 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
XZ3850000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

A8056 3-Amino-1,2,4-triazole, ≥95% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?