EMAIL THIS PAGE TO A FRIEND
A8732 Sigma-Aldrich

Acetoacetanilide

98%

  • CAS Number 102-01-2

  • Linear Formula CH3COCH2CONHC6H5

  • Molecular Weight 177.20

  •  Beilstein Registry Number 473419

  •  EC Number 202-996-4

  •  MDL number MFCD00008780

  •  PubChem Substance ID 24891360

Purchase

Properties

vapor density   6.1 (vs air)
InChI Key   DYRDKSSFIWVSNM-UHFFFAOYSA-N
assay   98%
autoignition temp.   843 °F
mp   83-88 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
Flash Point(F) 
323.6 °F
Flash Point(C) 
162 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?