EMAIL THIS PAGE TO A FRIEND
A93100 Aldrich

L-Aspartic acid

≥98%

Synonym: (S)-(+)-Aminosuccinic acid, (S)-Aminobutanedioic acid

  • CAS Number 56-84-8

  • Linear Formula HO2CCH2CH(NH2)CO2H

  • Molecular Weight 133.10

  •  Beilstein Registry Number 1723530

  •  EC Number 200-291-6

  •  MDL number MFCD00002616

  •  eCl@ss 32160406

  •  PubChem Substance ID 329771048

Purchase

Properties

InChI Key   CKLJMWTZIZZHCS-REOHCLBHSA-N
assay   ≥98%
optical activity   [α]20/D +25°, c = 8 in 5 M HCl
optical purity   ee: 98% (GLC)
mp   >300 °C (dec.)(lit.)

Description

Biochem/physiol Actions

Principal neurotransmitter for fast synaptic excitation.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
CI9098500

Documents

Certificate of Analysis

Certificate of Origin


Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?