• USA Home
  • B1251 - 6-Benzyloxy-5-methoxy-2-carboxyindole

EMAIL THIS PAGE TO A FRIEND
B1251 Sigma-Aldrich

6-Benzyloxy-5-methoxy-2-carboxyindole

≥99%, powder

Synonym: NSC 92530

  • CAS Number 2495-92-3

  • Empirical Formula (Hill Notation) C17H15NO4

  • Molecular Weight 297.31

  •  MDL number MFCD00055977

  •  PubChem Substance ID 57653916

Purchase

Properties

Related Categories Building Blocks, C13 to C27, Chemical Synthesis, Heterocyclic Building Blocks, Indoles More...
InChI Key   JMOOGCFXFNKYCS-UHFFFAOYSA-N
assay   ≥99%
form   powder
color   tan
storage temp.   −20°C
Gene Information   human ... SRD5A1(6715), SRD5A2(6716)

Description

Packaging

500 mg in poly bottle

Application

• Reactant for preparation of indolecarboxamides with antitumor activity in human lung cancer
• Reactant for preparation of benzophenone- and indolecarboxylic acids as potent type-2 specific inhibitors of human steroid 5α-reductase

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?