• USA Home
  • B23131 - 1-Benzyl-3-hydroxypyridinium chloride

EMAIL THIS PAGE TO A FRIEND
B23131 Sigma-Aldrich

1-Benzyl-3-hydroxypyridinium chloride

98%

  • CAS Number 3323-73-7

  • Empirical Formula (Hill Notation) C12H14ClNO

  • Molecular Weight 223.70

  •  MDL number MFCD00011990

Purchase

Properties

assay   98%
mp   162-164 °C (lit.)
SMILES string   [Cl-].Oc1ccc[n+](Cc2ccccc2)c1
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?