• USA Home
  • B3128 - S-Butyrylthiocholine chloride

EMAIL THIS PAGE TO A FRIEND
B3128 Sigma-Aldrich

S-Butyrylthiocholine chloride

crystalline

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds, Thiol Esters More...
form   crystalline
storage temp.   2-8°C
SMILES string   Cl.CCCC(=O)SCC[N](C)(C)C
InChI   1S/C9H20NOS.ClH/c1-5-6-9(11)12-8-7-10(2,3)4;/h5-8H2,1-4H3;1H/q+1;/p-1
InChI key   QYNZLUZQWOCGCH-UHFFFAOYSA-M

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?