• USA Home
  • B39607 - 1,1-Dichloro-2,2-bis(4-chlorophenyl)ethane

EMAIL THIS PAGE TO A FRIEND
B39607 Sigma-Aldrich

1,1-Dichloro-2,2-bis(4-chlorophenyl)ethane

technical grade

Synonym: 4,4′-DDD, TDE

  • CAS Number 72-54-8

  • Linear Formula (ClC6H4)2CHCHCl2

  • Molecular Weight 320.04

  •  Beilstein/REAXYS Number 1914072

  •  EC Number 200-783-0

  •  MDL number MFCD00000851

  •  PubChem Substance ID 24891743

Purchase

Properties

Related Categories Aryl, Building Blocks, C13 to C37+, Chemical Synthesis, Halogenated Hydrocarbons,
grade   technical grade
InChI Key   AHJKRLASYNVKDZ-UHFFFAOYSA-N
mp   94-96 °C
Gene Information   mouse ... Esr1(13982)
rat ... Ar(24208)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN2811 - class 6.1 - PG 3 - EHS - Toxic solids, organic, n.o.s., HI: all
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?