• USA Home
  • B44708 - 1,4-Bis(2-hydroxyethoxy)-2-butyne

EMAIL THIS PAGE TO A FRIEND
B44708 Sigma-Aldrich

1,4-Bis(2-hydroxyethoxy)-2-butyne

technical grade, 85%

Purchase

Properties

grade   technical grade
assay   85%
refractive index   n20/D 1.485 (lit.)
density   1.144 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Alkynes and Acetylenic Building Blocks

We list over 600 alkyne building blocks, including a wide array of propargyl alcohols, acetylenic boron reagents, and halogenated substrates for cross-coupling. Additionally, many of these are termin...
Keywords: Building blocks, Coupling reactions, Cross couplings

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?