• USA Home
  • B5003 - 10-Benzhydrylidene-9-anthrone

EMAIL THIS PAGE TO A FRIEND
B5003 Sigma-Aldrich

10-Benzhydrylidene-9-anthrone

Synonym: 9-Benzhydrylidene-10-anthrone

  • CAS Number 667-91-4

  • Empirical Formula (Hill Notation) C27H18O

  • Molecular Weight 358.43

  •  MDL number MFCD00019172

Purchase
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?