• USA Home
  • B89604 - 2-(tert-Butylamino)ethanol

EMAIL THIS PAGE TO A FRIEND
B89604 Sigma-Aldrich

2-(tert-Butylamino)ethanol

≥99%

Synonym: N-tert-Butylethanolamine

  • CAS Number 4620-70-6

  • Linear Formula (CH3)3CNHCH2CH2OH

  • Molecular Weight 117.19

  •  Beilstein/REAXYS Number 1732684

  •  EC Number 225-034-5

  •  MDL number MFCD00002837

  •  PubChem Substance ID 24892111

Purchase

Properties

vapor density   4 (vs air)
InChI Key   IUXYVKZUDNLISR-UHFFFAOYSA-N
assay   ≥99%
bp   90-92 °C/25 mmHg(lit.)
mp   41-43 °C (lit.)
density   0.867 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2923 6.1(8) / PGII
WGK Germany 
3
Flash Point(F) 
188.6 °F
Flash Point(C) 
87 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?