• USA Home
  • C118508 - Cyclopropyldiphenylmethanol

EMAIL THIS PAGE TO A FRIEND
C118508 Sigma-Aldrich

Cyclopropyldiphenylmethanol

97%

Purchase

Properties

assay   97%
mp   84-87 °C (lit.)
SMILES string   OC(C1CC1)(c2ccccc2)c3ccccc3
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?