• USA Home
  • C31406 - 3-Chlorocinnamic acid

EMAIL THIS PAGE TO A FRIEND
C31406 Sigma-Aldrich

3-Chlorocinnamic acid

98%

Synonym: trans-3-(3-Chlorophenyl)propenoic acid

  • CAS Number 1866-38-2

  • Linear Formula ClC6H4CH=CHCO2H

  • Molecular Weight 182.60

  •  Beilstein/REAXYS Number 2207378

  •  EC Number 217-478-3

  •  MDL number MFCD00004384

  •  PubChem Substance ID 24892566

Purchase

Properties

assay   98%
mp   161-164 °C (lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?