EMAIL THIS PAGE TO A FRIEND
C53 Sigma-Aldrich

Caffeine

99%

Synonym: 1,3,7-Trimethylxanthine

  • CAS Number 58-08-2

  • Empirical Formula (Hill Notation) C8H10N4O2

  • Molecular Weight 194.19

  •  Beilstein/REAXYS Number 17705

  •  EC Number 200-362-1

  •  MDL number MFCD00005758

Purchase

Properties

InChI Key   RYYVLZVUVIJVGH-UHFFFAOYSA-N
assay   99%
mp   234-236.5 °C (lit.)
Gene Information   human ... ADORA1(134), PDE4B(5142)

Description

Biochem/physiol Actions

A central nervous system stimulant believed to act through adenosine receptors and monoamine neurotransmitters. It is an adenosine receptor antagonist and adenosine 3′,5′-cyclic monophosphate (cAMP) phosphodiesterase inhibitor. Thus, levels of cAMP increase in cells following treatment with caffeine. It has been reported to affect cellular calcium levels, releasing calcium from intracellular stores. It overrides the cell cycle effects of various chemicals such as protease inhibitors, preventing apoptosis; and it has been shown to inhibit cellular DNA repair mechanisms.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
EV6475000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

C0750 Caffeine, powder, ReagentPlus®

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?