EMAIL THIS PAGE TO A FRIEND
C62816 Sigma-Aldrich

4-Chlorophenol

99%

  • CAS Number 106-48-9

  • Linear Formula ClC6H4OH

  • Molecular Weight 128.56

  •  Beilstein/REAXYS Number 507004

  •  EC Number 203-402-6

  •  MDL number MFCD00002318

Purchase

Properties

vapor pressure   1 mmHg ( 49.8 °C)
InChI Key   WXNZTHHGJRFXKQ-UHFFFAOYSA-N
assay   99%
bp   220 °C(lit.)
mp   42-45 °C
density   1.306 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2020 6.1 / PGIII
WGK Germany 
3
RTECS 
SK2800000
Flash Point(F) 
249.8 °F
Flash Point(C) 
121 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

185787 4-Chlorophenol, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?