• USA Home
  • CBR00278 - 1-[1-(1H-Imidazol-1-ylmethyl)cyclopropyl]methanamine

EMAIL THIS PAGE TO A FRIEND
CBR00278 Sigma-Aldrich

1-[1-(1H-Imidazol-1-ylmethyl)cyclopropyl]methanamine

AldrichCPR

Purchase

Properties

Related Categories Building Blocks, C3 to C8, Chemical Synthesis, Heterocyclic Building Blocks, Imidazoles More...
storage temp.   room temp
SMILES string   NCC1(CN2C=NC=C2)CC1
InChI   1S/C8H13N3/c9-5-8(1-2-8)6-11-4-3-10-7-11/h3-4,7H,1-2,5-6,9H2
InChI key   BMPMBBWPKSTTIE-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Legal Information

Product of ChemBridge Corp.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?