• USA Home
  • CBR00823 - 1-Isobutyl-4-methyl-1H-pyrazol-5-amine hydrochloride

EMAIL THIS PAGE TO A FRIEND
CBR00823 Sigma-Aldrich

1-Isobutyl-4-methyl-1H-pyrazol-5-amine hydrochloride

AldrichCPR

  • Empirical Formula (Hill Notation) C8H16ClN3

  • Molecular Weight 189.69

  •  MDL number MFCD13857375

  •  PubChem Substance ID 329776096

Purchase

Properties

Related Categories Building Blocks, C3-C9, C7 to C8, Chemical Synthesis, Halogenated Heterocycles,
SMILES string   NC(N(CC(C)C)N=C1)=C1C.[H]Cl
InChI   1S/C8H15N3.ClH/c1-6(2)5-11-8(9)7(3)4-10-11;/h4,6H,5,9H2,1-3H3;1H
InChI key   OOHCCXTXJFYFRB-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Legal Information

Product of ChemBridge Corp.

Safety & Documentation

Safety Information

RIDADR 
UN 2811 6.1 / PGIII

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?