• USA Home
  • CBR00917 - 3-(Aminomethyl)-N-methylbenzamide hydrochloride

EMAIL THIS PAGE TO A FRIEND
CBR00917 Sigma-Aldrich

3-(Aminomethyl)-N-methylbenzamide hydrochloride

AldrichCPR

  • Empirical Formula (Hill Notation) C9H13ClN2O

  • Molecular Weight 200.67

  •  MDL number MFCD09971582

  •  PubChem Substance ID 329776187

Purchase

Properties

Related Categories Amides, Amines, Building Blocks, C8 to C20+, C9,
SMILES string   O=C(NC)C1=CC=CC(CN)=C1.[H]Cl
InChI   1S/C9H12N2O.ClH/c1-11-9(12)8-4-2-3-7(5-8)6-10;/h2-5H,6,10H2,1H3,(H,11,12);1H
InChI key   VGPWKABXIGQSLA-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Legal Information

Product of ChemBridge Corp.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?