• USA Home
  • D150002 - 5,6-Dimethyl-1H-benzotriazole monohydrate

EMAIL THIS PAGE TO A FRIEND
D150002 Sigma-Aldrich

5,6-Dimethyl-1H-benzotriazole monohydrate

99%

  • CAS Number 1354973-50-4

  • Empirical Formula (Hill Notation) C8H9N3 · H2O

  • Molecular Weight 165.19

  •  Beilstein/REAXYS Number 122933

  •  EC Number 224-058-3

  •  MDL number MFCD00149344

  •  PubChem Substance ID 24893456

Purchase

Properties

assay   99%
mp   152-156 °C (lit.)
SMILES string   O.Cc1cc2nn[nH]c2cc1C
InChI   1S/C8H9N3.H2O/c1-5-3-7-8(4-6(5)2)10-11-9-7;/h3-4H,1-2H3,(H,9,10,11);1H2
InChI key   VVWPOQUENPOMAK-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles

Related Content

Indoles, Purines, and Their Isosteres

Substituted indoles and purines have frequently been referred to as “privileged structures” since they are capable of binding to multiple receptors with high affinity, and thus have applications acro...
Keywords: Building blocks, Medicinal chemistry

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?