EMAIL THIS PAGE TO A FRIEND
D174807 Sigma-Aldrich

2,5-Dimethylphenol

97%

Synonym: 2-Hydroxy-p-xylene, p-Xylenol

  • CAS Number 95-87-4

  • Linear Formula (CH3)2C6H3OH

  • Molecular Weight 122.16

  •  Beilstein/REAXYS Number 1099260

  •  EC Number 202-461-5

  •  MDL number MFCD00002237

Purchase

Properties

assay   97%
bp   212 °C (lit.)
mp   72-75 °C
SMILES string   Cc1ccc(C)c(O)c1
InChI   1S/C8H10O/c1-6-3-4-7(2)8(9)5-6/h3-5,9H,1-2H3
InChI key   NKTOLZVEWDHZMU-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2261 6.1 / PGII
WGK Germany 
3
RTECS 
ZE5775000
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

D174602 2,5-Dimethylphenol, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?