• USA Home
  • D21804 - 4,6-Diamino-5-nitropyrimidine

EMAIL THIS PAGE TO A FRIEND
D21804 Aldrich

4,6-Diamino-5-nitropyrimidine

  • CAS Number 2164-84-3

  • Empirical Formula (Hill Notation) C4H5N5O2

  • Molecular Weight 155.11

  •  MDL number MFCD00023268

Purchase
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?