EMAIL THIS PAGE TO A FRIEND
D99706 Sigma-Aldrich

Diethyl pimelate

97%

Synonym: Diethyl heptanedioate, Pimelic acid diethyl ester

  • CAS Number 2050-20-6

  • Linear Formula C2H5OCO(CH2)5COOC2H5

  • Molecular Weight 216.27

  •  Beilstein/REAXYS Number 1710126

  •  EC Number 218-083-9

  •  MDL number MFCD00009216

  •  PubChem Substance ID 24894354

Purchase

Properties

Related Categories Building Blocks, C10 to C11, Carbonyl Compounds, Chemical Synthesis, Esters,
assay   97%
refractive index   n20/D 1.430 (lit.)
bp   192-194 °C/100 mmHg (lit.)
density   0.994 g/mL at 25 °C (lit.)
SMILES string   CCOC(=O)CCCCCC(=O)OCC
InChI   1S/C11H20O4/c1-3-14-10(12)8-6-5-7-9-11(13)15-4-2/h3-9H2,1-2H3
InChI key   LKKOGZVQGQUVHF-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?