• USA Home
  • E004 - 3-Hydroxymethyl-β-carboline

EMAIL THIS PAGE TO A FRIEND
E004 Sigma-Aldrich

3-Hydroxymethyl-β-carboline

Synonym: 3-HMC

  • CAS Number 65474-79-5

  • Empirical Formula (Hill Notation) C12H10N2O

  • Molecular Weight 198.22

  •  MDL number MFCD00055074

  •  PubChem Substance ID 329798936

  •  NACRES NA.22

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, N-Containing, Other Heterocyclic Building Blocks More...
mp   215-219 °C
SMILES string   OCc1cc2c(cn1)[nH]c3ccccc23
InChI   1S/C12H10N2O/c15-7-8-5-10-9-3-1-2-4-11(9)14-12(10)6-13-8/h1-6,14-15H,7H2
InChI key   CPBYHTDUBNSBQM-UHFFFAOYSA-N

Description

Packaging

100 mg in poly bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?