EMAIL THIS PAGE TO A FRIEND
E304 Sigma-Aldrich

Elaidic acid

98%

Synonym: trans-9-Octadecenoic acid, trans-Oleic acid

  • CAS Number 112-79-8

  • Linear Formula CH3(CH2)7CH=CH(CH2)7COOH

  • Molecular Weight 282.46

  •  Beilstein/REAXYS Number 1726543

  •  EC Number 204-006-6

  •  MDL number MFCD00063954

Purchase

Properties

assay   98%
bp   288 °C/100 mmHg (lit.)
mp   42-44 °C (lit.)
SMILES string   CCCCCCCC\C=C\CCCCCCCC(O)=O
InChI   1S/C18H34O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h9-10H,2-8,11-17H2,1H3,(H,19,20)/b10-9+
InChI key   ZQPPMHVWECSIRJ-MDZDMXLPSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
JX6125000
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

E4637 Elaidic acid, ≥99.0% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?