EMAIL THIS PAGE TO A FRIEND
E51228 Sigma-Aldrich

N-Ethylurethane

99%

Synonym: Ethyl N-ethylcarbamate

Purchase

Properties

assay   99%
refractive index   n20/D 1.421 (lit.)
bp   85 °C/20 mmHg(lit.)
density   0.981 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993 / PGIII
WGK Germany 
3
RTECS 
FA9100000
Flash Point(F) 
168.8 °F
Flash Point(C) 
76 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?