• USA Home
  • H36389 - 5-Benzoyl-4-hydroxy-2-methoxybenzenesulfonic acid

EMAIL THIS PAGE TO A FRIEND
H36389 Sigma-Aldrich

5-Benzoyl-4-hydroxy-2-methoxybenzenesulfonic acid

97%

Synonym: 2-Hydroxy-4-methoxybenzophenone-5-sulfonic acid, HMBS, Sulisobenzone

  • CAS Number 4065-45-6

  • Empirical Formula (Hill Notation) C14H12O6S

  • Molecular Weight 308.31

  •  Beilstein/REAXYS Number 2889165

  •  EC Number 223-772-2

  •  MDL number MFCD00024962

Purchase

Properties

assay   97%
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?