EMAIL THIS PAGE TO A FRIEND
H4625 Sigma-Aldrich

Hexachlorophene

crystalline

Synonym: 2,2′-Methylenebis(3,4,6-trichlorophenol)

  • CAS Number 70-30-4

  • Linear Formula CH2[C6H(Cl)3OH]2

  • Molecular Weight 406.90

  •  Beilstein/REAXYS Number 2064407

  •  EC Number 200-733-8

  •  MDL number MFCD00002171

  •  PubChem Substance ID 24895645

Purchase

Properties

form   crystalline
color   off-white to tan
mp   163-165 °C (lit.)
SMILES string   Oc1c(Cl)cc(Cl)c(Cl)c1Cc2c(O)c(Cl)cc(Cl)c2Cl
InChI   1S/C13H6Cl6O2/c14-6-2-8(16)12(20)4(10(6)18)1-5-11(19)7(15)3-9(17)13(5)21/h2-3,20-21H,1H2
InChI key   ACGUYXCXAPNIKK-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2875 6.1 / PGIII
WGK Germany 
3
RTECS 
SM0700000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?