• USA Home
  • H59117 - 6-Hydroxytetrahydropyran-2-carboxylic acid lactone

EMAIL THIS PAGE TO A FRIEND
H59117 Sigma-Aldrich

6-Hydroxytetrahydropyran-2-carboxylic acid lactone

Synonym: 6,8-Dioxabicyclo[3.2.1]octan-7-one

  • CAS Number 5257-20-5

  • Empirical Formula (Hill Notation) C6H8O3

  • Molecular Weight 128.13

  •  MDL number MFCD00168003

Purchase

Properties

SMILES string   O=C1O[C@H]2CCC[C@@H]1O2
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?