EMAIL THIS PAGE TO A FRIEND
H7009 Sigma-Aldrich

1-Hexadecene

technical grade, 92%

Synonym: Cetene

  • CAS Number 629-73-2

  • Linear Formula CH3(CH2)13CH=CH2

  • Molecular Weight 224.43

  •  Beilstein/REAXYS Number 1753401

  •  EC Number 211-105-8

  •  MDL number MFCD00008991

  •  PubChem Substance ID 329815238

Purchase

Properties

Related Categories Acyclic, Alkenes, Building Blocks, Chemical Synthesis, Organic Building Blocks More...
grade   technical grade
vapor density   >0.8 (vs air)
vapor pressure   <0.2 mmHg ( 20 °C)
InChI Key   GQEZCXVZFLOKMC-UHFFFAOYSA-N
assay   92%
autoignition temp.   464 °F
impurities   ≤2.0% 2-butyl-1-dodecene
  ≤2.0% 2-ethyl-1-tetradecene
  ≤2.0% 2-hexyl-1-decene
  ≤2.0% n-hexadecane
refractive index   n20/D 1.441 (lit.)
bp   274 °C (lit.)
mp   3-5 °C (lit.)
density   0.783 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
MM0479000
Flash Point(F) 
269.6 °F
Flash Point(C) 
132 °C

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?