• USA Home
  • J100037 - Tantalum(V) ethoxide

EMAIL THIS PAGE TO A FRIEND
J100037 Sigma-Aldrich

Tantalum(V) ethoxide

electronic grade, ≥95%, ≥99.999% metals basis

Synonym: Pentaethoxytantalum, Tantalum pentaethoxide

  • CAS Number 6074-84-6

  • Linear Formula Ta(OC2H5)5

  • Molecular Weight 406.25

  •  Beilstein/REAXYS Number 3678999

  •  EC Number 228-010-2

  •  MDL number MFCD00049785

Purchase

Properties

grade   electronic grade
description   J100037
assay   ≥95%
  ≥99.999% metals basis
form   liquid
refractive index   n20/D 1.487 (lit.)
bp   155 °C/0.01 mmHg (lit.)
  155 °C/0.01 mmHg
mp   21 °C (lit.)
  21 °C
density   1.566 g/mL at 25 °C (lit.)
  1.566 g/mL at 25 °C
SMILES string   CCO[Ta](OCC)(OCC)(OCC)OCC
InChI   1S/5C2H5O.Ta/c5*1-2-3;/h5*2H2,1H3;/q5*-1;+5
InChI key   HSXKFDGTKKAEHL-UHFFFAOYSA-N

Description

General description

Please inquire for quantity, pricing, and packaging options.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2920 3(8) / PGII
WGK Germany 
3
Flash Point(F) 
86 °F
Flash Point(C) 
30 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?