• USA Home
  • L500542 - 6-Oxa-1-azaspiro[3.3]heptane

EMAIL THIS PAGE TO A FRIEND
L500542 Sigma-Aldrich

6-Oxa-1-azaspiro[3.3]heptane

AldrichCPR

  • Empirical Formula (Hill Notation) C5H9NO

  • Molecular Weight 99.13

  •  MDL number MFCD13180595

Purchase

Properties

Quality Level   100
availability   not available in EU, Japan
SMILES string   C1CC2(COC2)N1

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Sales restrictions may apply.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Oxetanes

Oxetanes are the closest homologs to epoxides, but historically have received far less attention than their three-membered ringed brethren. However, oxetanes have received increasing exposure as attr...
Mark Redlich
Chemfiles Volume 10 Article 4
Keywords: Antibiotics, Building blocks, Drug discovery, Medicinal chemistry

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?