EMAIL THIS PAGE TO A FRIEND
L754099 Sigma-Aldrich

diF-TES-ADT

AldrichCPR, >99%

Synonym: 2,8-Difluoro-5,11-bis(triethylsilylethynyl)anthradithiophene

  • CAS Number 1015071-21-2

  • Empirical Formula (Hill Notation) C34H36F2S2Si2

  • Molecular Weight 602.95

Purchase

Properties

assay   >99%
loss   0.5 wt. %, 250 °C
λmax   302 nm in methylene chloride
SMILES string   CC[Si](CC)(CC)C#Cc1c2cc3cc(F)sc3cc2c(C#C[Si](CC)(CC)CC)c4cc5cc(F)sc5cc14

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?