EMAIL THIS PAGE TO A FRIEND
M1210 Sigma-Aldrich

DL-Malic acid

ReagentPlus®, 99%

Synonym: (±)-2-Hydroxysuccinic acid, DL-Hydroxybutanedioic acid

  • CAS Number 6915-15-7

  • Linear Formula HO2CCH2CH(OH)CO2H

  • Molecular Weight 134.09

  •  Beilstein/REAXYS Number 1723539

  •  EC Number 230-022-8

  •  MDL number MFCD00064212

  •  PubChem Substance ID 57654350

Purchase

Properties

Related Categories Building Blocks, C1 to C5, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
grade   ReagentPlus®
vapor density   4.6 (vs air)
vapor pressure   <0.1 mmHg ( 20 °C)
InChI Key   BJEPYKJPYRNKOW-UHFFFAOYSA-N
assay   99%
autoignition temp.   644 °F
mp   131-133 °C (lit.)

Description

Legal Information

ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
ON7175000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?