EMAIL THIS PAGE TO A FRIEND
M80407 Sigma-Aldrich

2-Methylresorcinol

technical grade, 90%

Synonym: 2,6-Dihydroxytoluene

  • CAS Number 608-25-3

  • Linear Formula CH3C6H3(OH)2

  • Molecular Weight 124.14

  •  Beilstein/REAXYS Number 2042177

  •  EC Number 210-155-8

  •  MDL number MFCD00002271

  •  PubChem Substance ID 24897216

  •  NACRES NA.22

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds, Polyols More...
Quality Level   200
grade   technical grade
assay   90%
bp   264 °C (lit.)
mp   114-120 °C (lit.)
SMILES string   Cc1c(O)cccc1O
InChI   1S/C7H8O2/c1-5-6(8)3-2-4-7(5)9/h2-4,8-9H,1H3
InChI key   ZTMADXFOCUXMJE-UHFFFAOYSA-N

Description

Packaging

100, 500 g in poly bottle

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
VH2009500
Flash Point(F) 
275 °F
Flash Point(C) 
135 °C

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles

Articles

HPLC Analysis of Catechols and Resorcinols on Ascentis® RP-Amide

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Catechols and Resorcinols on Ascentis® RP-Amide
Keywords: Chromatography, Clinical, High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?