EMAIL THIS PAGE TO A FRIEND
N18501 Sigma-Aldrich

2-Nitromesitylene

99%

Synonym: 1,3,5-Trimethyl-2-nitrobenzene

Purchase

Properties

assay   99%
bp   255 °C (lit.)
mp   41-44 °C (lit.)
SMILES string   Cc1cc(C)c(c(C)c1)[N+]([O-])=O
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
186.8 °F
Flash Point(C) 
86 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?