EMAIL THIS PAGE TO A FRIEND
O7600 Sigma-Aldrich

Oleyl alcohol

technical grade, 65%

Synonym: Z-9-Dodecen-1-ol, cis-9-Octadecen-1-ol

  • CAS Number 143-28-2

  • Linear Formula CH3(CH2)7CH=CH(CH2)7CH2OH

  • Molecular Weight 268.48

  •  Beilstein/REAXYS Number 1723962

  •  EC Number 205-597-3

  •  MDL number MFCD00002993

Purchase

Properties

grade   technical grade
assay   65%
refractive index   n20/D 1.46 (lit.)
bp   207 °C/13 mmHg (lit.)
mp   0-5.0 °C (lit.)
density   0.849 g/mL at 25 °C (lit.)
SMILES string   CCCCCCCC\C=C/CCCCCCCCO
InChI   1S/C18H36O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h9-10,19H,2-8,11-18H2,1H3/b10-9-
InChI key   ALSTYHKOOCGGFT-KTKRTIGZSA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
RG4120000
Flash Point(F) 
338 °F
Flash Point(C) 
170 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?