EMAIL THIS PAGE TO A FRIEND
Q1875 Sigma-Aldrich

3-Quinuclidinol

97%

Synonym: 1-Azabicyclo[2.2.2]octan-3-ol, 3-Hydroxyquinuclidine

  • CAS Number 1619-34-7

  • Empirical Formula (Hill Notation) C7H13NO

  • Molecular Weight 127.18

  •  Beilstein/REAXYS Number 80122

  •  EC Number 216-578-4

  •  MDL number MFCD00151326

Purchase

Properties

assay   97%
mp   220-223 °C (lit.)
SMILES string   OC1CN2CC[C@H]1CC2
InChI   1S/C7H13NO/c9-7-5-8-3-1-6(7)2-4-8/h6-7,9H,1-5H2
InChI key   IVLICPVPXWEGCA-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3263 8 / PGII
WGK Germany 
3
RTECS 
VD6191700
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

253340 3-Quinuclidinol, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?