• USA Home
  • S2805 - 5-Hydroxytryptamine creatinine sulfate complex

EMAIL THIS PAGE TO A FRIEND
S2805 Sigma-Aldrich

5-Hydroxytryptamine creatinine sulfate complex

99%

Synonym: 5-Hydroxytryptamine creatinine sulfate salt

  • CAS Number 61-47-2

  • Empirical Formula (Hill Notation) C10H12N2O

  • Molecular Weight 176.22

  •  MDL number MFCD00149653

Purchase

Properties

assay   99%
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?