• USA Home
  • T78700 - 2,3,6-Trimethylphenol

EMAIL THIS PAGE TO A FRIEND
T78700 Sigma-Aldrich

2,3,6-Trimethylphenol

95%

Purchase

Properties

Related Categories Building Blocks, C9 to C20+, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds,
InChI Key   QQOMQLYQAXGHSU-UHFFFAOYSA-N
assay   95%
mp   59-62 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2430 8 / PGIII
WGK Germany 
1
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?