EMAIL THIS PAGE TO A FRIEND
U1206 Sigma-Aldrich

2-Undecanol

97%

Synonym: Methyl nonyl carbinol

  • CAS Number 1653-30-1

  • Linear Formula CH3(CH2)8CH(OH)CH3

  • Molecular Weight 172.31

  •  Beilstein/REAXYS Number 1719795

  •  EC Number 216-722-6

  •  MDL number MFCD00021958

Purchase

Properties

InChI Key   XMUJIPOFTAHSOK-UHFFFAOYSA-N
assay   97%
refractive index   n20/D 1.437 (lit.)
bp   130-131 °C/28 mmHg (lit.)
  229-231 °C (lit.)
mp   2-3 °C
density   0.828 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
2
Flash Point(F) 
206.6 °F
Flash Point(C) 
97 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

94070 2-Undecanol, ≥98.0% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?