EMAIL THIS PAGE TO A FRIEND
W209624 Sigma-Aldrich

Anise star oil

Certified organic (NOP)

Purchase

Properties

Related Categories A-B, A-D, Alphabetical Listings, Certified Organics, Essential Oils,
biological source   Illicium verum Hook. f. (also Badiane)
grade   Certified organic (NOP)
  Kosher
  follows IFRA guidelines
reg. compliance   EU Regulation 1223/2009
  FDA 21 CFR (117)
optical activity   [α]20/D −1°, neat
composition   may contain Limonene (sum of D, L and DL),Linalool and EU 1223/2009 restricted Methyl Chavicol, Anisyl Alcohol, Anisaldehyde
origin   Vietnam origin
refractive index   n20/D 1.554(lit.)
bp   232 °C(lit.)
density   0.980 g/mL at 25 °C(lit.)
Documentation   see Safety & Documentation for available documents
Organoleptic   anise
food allergen   no known allergens
fragrance allergen   anise alcohol, linalool, limonene (sum of D, L and DL)

Description

Packaging

1 kg in glass bottle

Legal Information

Certified organic by MOSA

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN3082 - class 9 - PG 3 - DOT NA1993 - Environmentally hazardous substances, liquid, n.o.s. HI: all (not BR)
WGK Germany 
3
Flash Point(F) 
197.6 °F
Flash Point(C) 
92 °C
Protocols & Articles

Articles

Understanding the Complexities of Kosher Ingredients

Introduction What is Kosher Product Considerations for Kosher Certification Kosher's Added Benefit to the Food Industry Sigma-Aldrich's Kosher Experience and Offering
Dr. Luke Grocholl1, Rabbi Gershon Segal

1Dr. Luke Grocholl, Quality Assurance Supervisor, Sigma-Aldrich Flavors & Fragrances
Keywords: Fermentation, Food & Beverage, Safety industry

Related Content

Flavors and Fragrances Quality and Regulatory

From a comprehensive vendor approval process, to the testing of raw material at receipt as well as when product is ready to be shipped; we prove our dedication to supply chain transparency, every day.
Keywords: Allergens, Food Safety, Solvents

Related Products

related product

Product #

Description

Add to Cart

07475 Timestrip® Food
06732 Timestrip® Seafood

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?